The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002773.4 | NP_002764.1 | 3 - 4 | 528 | 60 |
GenBank mRNA | AU142128.1 | - | 3 - 4 | 528 | 60 |
BC001462.2 | - | 3 - 4 | 439 | 60 | |
L41351.1 | - | 3 - 4 | 492 | 60 |