Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
Chromosome Location
Chr.3: 53071151 - 53130469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052859.3 NP_443091.1 6 - 7 745 96
XM_005265537.4 XP_005265594.1 6 - 7 758 96
XM_006713384.3 XP_006713447.1 6 - 7 756 96
XM_011534214.2 XP_011532516.1 6 - 7 760 96
XM_011534215.2 XP_011532517.1 6 - 7 760 96
XM_017007460.1 XP_016862949.1 6 - 7 757 96
GenBank mRNA AJ318099.1 - 6 - 7 729 96
BC043595.1 - 6 - 7 738 96

View Full Product Details