Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
Chromosome Location
Chr.3: 53071151 - 53130469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052859.3 NP_443091.1 7 - 8 832 119
XM_005265537.4 XP_005265594.1 7 - 8 845 119
XM_006713384.3 XP_006713447.1 7 - 8 843 119
XM_011534214.2 XP_011532516.1 7 - 8 847 119
XM_011534215.2 XP_011532517.1 7 - 8 847 119
XM_011534216.2 XP_011532518.1 7 - 8 930 119
XM_017007460.1 XP_016862949.1 7 - 8 844 119
XM_017007461.1 XP_016862950.1 8 - 9 1016 119
GenBank mRNA AJ318099.1 - 7 - 8 816 119
BC043595.1 - 7 - 8 825 119

View Full Product Details