Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
Chromosome Location
Chr.3: 53071151 - 53130469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052859.3 NP_443091.1 8 - 9 880 90
XM_005265537.4 XP_005265594.1 8 - 9 893 90
XM_006713384.3 XP_006713447.1 8 - 9 891 90
XM_011534214.2 XP_011532516.1 8 - 9 895 90
XM_011534215.2 XP_011532517.1 8 - 9 895 90
XM_011534216.2 XP_011532518.1 8 - 9 978 90
XM_017007460.1 XP_016862949.1 8 - 9 892 90
XM_017007461.1 XP_016862950.1 9 - 10 1064 90
GenBank mRNA AJ318099.1 - 8 - 9 864 90
BC043595.1 - 8 - 9 873 90

View Full Product Details