The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017770.3 | NP_060240.3 | 5 - 6 | 661 | 68 |
XM_011514716.2 | XP_011513018.1 | 5 - 6 | 613 | 68 | |
XM_011514717.2 | XP_011513019.1 | 5 - 6 | 579 | 68 | |
GenBank mRNA | BC050278.1 | - | 5 - 6 | 661 | 68 |
BC060809.1 | - | 5 - 6 | 612 | 68 |