Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGI1
Entrez Gene ID
9223
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location
Chr.3: 65353525 - 66038836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033057.1 NP_001028229.1 4 - 5 1282 97
NM_004742.2 NP_004733.2 4 - 5 1282 97
NM_015520.1 NP_056335.1 4 - 5 1282 97
XM_005265564.1 XP_005265621.1 4 - 5 1284 97
XM_005265566.1 XP_005265623.1 4 - 5 1284 97
XM_005265568.1 XP_005265625.1 4 - 5 1284 97
XM_005265571.1 XP_005265628.1 4 - 5 1284 97
XM_005265574.3 XP_005265631.1 4 - 5 1284 97
XM_006713408.1 XP_006713471.1 4 - 5 1284 97
XM_006713410.1 XP_006713473.1 4 - 5 1284 97
XM_006713412.1 XP_006713475.1 4 - 5 1284 97
XM_017007498.1 XP_016862987.1 4 - 5 1284 97
XM_017007500.1 XP_016862989.1 4 - 5 1638 97
XM_017007504.1 XP_016862993.1 4 - 5 1284 97
XM_017007506.1 XP_016862995.1 4 - 5 1284 97
XM_017007507.1 XP_016862996.1 3 - 4 448 97
XM_017007508.1 XP_016862997.1 3 - 4 448 97
GenBank mRNA AB010894.1 - 4 - 5 998 97
AF401654.1 - 4 - 5 756 97
AF401655.1 - 4 - 5 756 97
AF401656.1 - 4 - 5 756 97
AK289803.1 - 4 - 5 851 97
KC170987.1 - 4 - 5 756 97
U80754.1 - 2 - 3 309 97
U96115.1 - 2 - 3 303 97

View Full Product Details