Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
Chromosome Location
Chr.5: 43486699 - 43515433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198566.3 NP_940968.1 12 - 13 2119 141
XM_017009445.1 XP_016864934.1 11 - 12 2150 141
XM_017009446.1 XP_016864935.1 10 - 11 2060 141
GenBank mRNA AK056925.1 - 12 - 13 2034 141
BC036867.1 - 12 - 13 1982 141

View Full Product Details