Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
Chromosome Location
Chr.5: 43486699 - 43515433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198566.3 NP_940968.1 9 - 10 1884 154
XM_011514038.2 XP_011512340.1 9 - 10 1793 154
XM_017009445.1 XP_016864934.1 8 - 9 1915 154
XM_017009446.1 XP_016864935.1 7 - 8 1825 154
GenBank mRNA AK056925.1 - 9 - 10 1799 154
BC036867.1 - 9 - 10 1747 154

View Full Product Details