The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031948.4 | NP_114154.1 | 2 - 3 | 466 | 75 |
GenBank mRNA | AB056161.1 | - | 2 - 3 | 125 | 75 |
AJ306593.1 | - | 2 - 3 | 94 | 75 | |
AY030095.1 | - | 2 - 3 | 75 | 75 | |
AY359106.1 | - | 2 - 3 | 110 | 75 | |
BC034294.2 | - | 2 - 3 | 130 | 75 |