The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031948.4 | NP_114154.1 | 3 - 4 | 627 | 137 |
GenBank mRNA | AB056161.1 | - | 3 - 4 | 286 | 137 |
AJ306593.1 | - | 3 - 4 | 255 | 137 | |
AY030095.1 | - | 3 - 4 | 236 | 137 | |
AY359106.1 | - | 3 - 4 | 271 | 137 | |
BC034294.2 | - | 3 - 4 | 291 | 137 |