The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318395.1 | NP_001305324.1 | 3 - 4 | 734 | 85 |
NM_031948.4 | NP_114154.1 | 4 - 5 | 897 | 85 | |
XM_011522689.2 | XP_011520991.1 | 2 - 3 | 626 | 85 | |
GenBank mRNA | AB056161.1 | - | 4 - 5 | 556 | 85 |
AJ306593.1 | - | 4 - 5 | 525 | 85 | |
AK055576.1 | - | 1 - 2 | 1498 | 85 | |
AK310214.1 | - | 3 - 4 | 409 | 85 | |
AY030095.1 | - | 4 - 5 | 506 | 85 | |
AY359106.1 | - | 4 - 5 | 541 | 85 | |
BC034294.2 | - | 4 - 5 | 561 | 85 | |
BC036912.2 | - | 3 - 4 | 745 | 85 |