The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146688.1 | NP_001140160.1 | 15 - 16 | 2378 | 67 |
NM_018433.5 | NP_060903.2 | 15 - 16 | 2647 | 67 | |
XM_006712051.3 | XP_006712114.1 | 14 - 15 | 2345 | 67 | |
XM_017004493.1 | XP_016859982.1 | 15 - 16 | 2666 | 67 | |
XM_017004494.1 | XP_016859983.1 | 16 - 17 | 2489 | 67 | |
GenBank mRNA | AB018285.2 | - | 15 - 16 | 2410 | 67 |
AK074314.1 | - | 1 - 2 | 44 | 67 | |
AK304276.1 | - | 13 - 14 | 2164 | 67 | |
BC038297.2 | - | 15 - 16 | 2424 | 67 | |
BX640698.1 | - | 15 - 16 | 2448 | 67 | |
BX647752.1 | - | 15 - 16 | 2476 | 67 | |
CR749581.1 | - | 15 - 16 | 2415 | 67 |