The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146688.1 | NP_001140160.1 | 3 - 4 | 402 | 100 |
NM_018433.5 | NP_060903.2 | 3 - 4 | 671 | 100 | |
XM_006712051.3 | XP_006712114.1 | 2 - 3 | 374 | 100 | |
XM_017004493.1 | XP_016859982.1 | 3 - 4 | 690 | 100 | |
XM_017004494.1 | XP_016859983.1 | 3 - 4 | 460 | 100 | |
GenBank mRNA | AB018285.2 | - | 3 - 4 | 434 | 100 |
AL832150.1 | - | 3 - 4 | 437 | 100 | |
BC038297.2 | - | 3 - 4 | 448 | 100 | |
BG723267.1 | - | 3 - 4 | 453 | 100 | |
BX640698.1 | - | 3 - 4 | 472 | 100 | |
BX647271.1 | - | 3 - 4 | 440 | 100 | |
BX647752.1 | - | 3 - 4 | 500 | 100 | |
BX648841.1 | - | 3 - 4 | 403 | 100 | |
BX649109.1 | - | 3 - 4 | 441 | 100 |