The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001227.4 | NP_001218.1 | 3 - 4 | 557 | 88 |
NM_001267056.1 | NP_001253985.1 | 3 - 4 | 435 | 88 | |
NM_001267057.1 | NP_001253986.1 | 3 - 4 | 588 | 88 | |
NM_001267058.1 | NP_001253987.1 | 2 - 3 | 239 | 88 | |
NM_001320911.1 | NP_001307840.1 | 3 - 4 | 466 | 88 | |
NM_033338.5 | NP_203124.1 | 4 - 5 | 664 | 88 | |
NM_033339.4 | NP_203125.1 | 4 - 5 | 631 | 88 | |
NM_033340.3 | NP_203126.1 | 3 - 4 | 435 | 88 | |
XM_006718017.3 | XP_006718080.1 | 4 - 5 | 368 | 88 | |
XM_011540260.1 | XP_011538562.1 | 2 - 3 | 587 | 88 | |
XM_017016763.1 | XP_016872252.1 | 3 - 4 | 615 | 88 | |
XM_017016764.1 | XP_016872253.1 | 3 - 4 | 382 | 88 | |
GenBank mRNA | AB451281.1 | - | 3 - 4 | 346 | 88 |
AB451413.1 | - | 3 - 4 | 346 | 88 | |
AK223489.1 | - | 3 - 4 | 328 | 88 | |
AK298964.1 | - | 2 - 3 | 200 | 88 | |
AK300578.1 | - | 3 - 4 | 314 | 88 | |
AU130224.1 | - | 3 - 4 | 349 | 88 | |
BC015799.1 | - | 3 - 4 | 325 | 88 | |
BT006683.1 | - | 2 - 3 | 247 | 88 | |
BX487250.1 | - | 3 - 4 | 423 | 88 | |
BX647973.1 | - | 3 - 4 | 366 | 88 | |
DC364916.1 | - | 3 - 4 | 436 | 88 | |
DC375817.1 | - | 3 - 4 | 438 | 88 | |
DC384804.1 | - | 4 - 5 | 413 | 88 | |
DC405356.1 | - | 3 - 4 | 393 | 88 | |
DC407017.1 | - | 3 - 4 | 464 | 88 | |
U37448.1 | - | 3 - 4 | 290 | 88 | |
U37449.1 | - | 3 - 4 | 393 | 88 | |
U39613.1 | - | 2 - 3 | 247 | 88 | |
U40281.1 | - | 3 - 4 | 294 | 88 | |
U67206.1 | - | 4 - 5 | 633 | 88 | |
U67320.1 | - | 3 - 4 | 286 | 88 |