The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177.5 | NP_001168.1 | 3 - 4 | 400 | 98 |
NM_001301068.1 | NP_001287997.1 | 2 - 3 | 262 | 98 | |
GenBank mRNA | AF493887.1 | - | 2 - 3 | 225 | 98 |
AK299062.1 | - | 3 - 4 | 337 | 98 | |
AK301701.1 | - | 3 - 4 | 305 | 98 | |
AK302911.1 | - | 2 - 3 | 188 | 98 | |
AK311793.1 | - | 3 - 4 | 325 | 98 | |
AL701988.1 | - | 3 - 4 | 371 | 98 | |
BC007000.1 | - | 3 - 4 | 355 | 98 | |
BT007260.1 | - | 2 - 3 | 225 | 98 | |
BX537387.1 | - | 3 - 4 | 372 | 98 | |
CD686919.1 | - | 3 - 4 | 405 | 98 | |
DC343685.1 | - | 3 - 4 | 400 | 98 | |
L28997.1 | - | 3 - 4 | 369 | 98 |