The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015877.1 | NP_001015877.1 | 10 - 11 | 1297 | 161 |
GenBank mRNA | AB058726.1 | - | 10 - 11 | 1183 | 161 |
AK303369.1 | - | 9 - 10 | 1160 | 161 | |
CF242973.1 | - | 4 - 5 | 436 | 161 |