The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015877.1 | NP_001015877.1 | 2 - 3 | 342 | 124 |
NM_032335.3 | NP_115711.2 | 2 - 3 | 342 | 124 | |
NM_032458.2 | NP_115834.1 | 2 - 3 | 342 | 124 | |
GenBank mRNA | AB058726.1 | - | 2 - 3 | 228 | 124 |
AK290095.1 | - | 2 - 3 | 313 | 124 | |
AL832725.1 | - | 2 - 3 | 335 | 124 | |
AY157622.1 | - | 2 - 3 | 342 | 124 | |
BC005994.1 | - | 2 - 3 | 281 | 124 | |
CD685336.1 | - | 2 - 3 | 350 | 124 | |
DR000282.1 | - | 2 - 3 | 377 | 124 |