The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160167.1 | NP_001153639.1 | 5 - 6 | 738 | 79 |
NM_001160168.1 | NP_001153640.1 | 3 - 4 | 278 | 79 | |
NM_001160169.1 | NP_001153641.1 | 4 - 5 | 474 | 79 | |
NM_024841.4 | NP_079117.3 | 6 - 7 | 724 | 79 | |
GenBank mRNA | AK024275.1 | - | 3 - 4 | 225 | 79 |
AK055042.1 | - | 5 - 6 | 625 | 79 | |
BX640959.1 | - | 6 - 7 | 705 | 79 | |
DR001466.1 | - | 4 - 5 | 407 | 79 |