Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
Chromosome Location
Chr.15: 51447724 - 51622833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174116.1 NP_001167587.1 23 - 24 5709 98
NM_001174117.1 NP_001167588.1 21 - 22 3801 98
NM_015263.3 NP_056078.2 23 - 24 5709 98
XM_005254255.1 XP_005254312.1 23 - 24 5772 98
XM_005254256.1 XP_005254313.1 23 - 24 5772 98
XM_017022034.1 XP_016877523.1 23 - 24 5445 98
GenBank mRNA AB020663.2 - 11 - 12 3086 98
AF389880.1 - 23 - 24 5691 98
AK123284.1 - 6 - 7 1536 98
BC140781.1 - 21 - 22 3666 98
BC144539.1 - 23 - 24 5574 98

View Full Product Details