Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
Chromosome Location
Chr.15: 51447724 - 51622833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174116.1 NP_001167587.1 37 - 38 8549 120
NM_001174117.1 NP_001167588.1 35 - 36 6638 120
NM_015263.3 NP_056078.2 37 - 38 8546 120
XM_005254255.1 XP_005254312.1 38 - 39 8675 120
XM_005254256.1 XP_005254313.1 38 - 39 8672 120
XM_017022034.1 XP_016877523.1 38 - 39 8348 120
GenBank mRNA AB020663.2 - 25 - 26 5926 120
AF389880.1 - 37 - 38 8528 120
BC016032.1 - 15 - 16 2761 120
BC022050.1 - 8 - 9 866 120
BC140781.1 - 35 - 36 6503 120
BC144539.1 - 37 - 38 8414 120

View Full Product Details