The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174116.1 | NP_001167587.1 | 42 - 43 | 9134 | 92 |
NM_001174117.1 | NP_001167588.1 | 40 - 41 | 7223 | 92 | |
NM_015263.3 | NP_056078.2 | 42 - 43 | 9131 | 92 | |
XM_005254255.1 | XP_005254312.1 | 43 - 44 | 9260 | 92 | |
XM_005254256.1 | XP_005254313.1 | 43 - 44 | 9257 | 92 | |
XM_017022034.1 | XP_016877523.1 | 43 - 44 | 8933 | 92 | |
GenBank mRNA | AB020663.2 | - | 30 - 31 | 6511 | 92 |
BC016032.1 | - | 20 - 21 | 3346 | 92 | |
BC022050.1 | - | 13 - 14 | 1451 | 92 | |
BC140781.1 | - | 40 - 41 | 7088 | 92 | |
BC144539.1 | - | 42 - 43 | 8999 | 92 |