The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174116.1 | NP_001167587.1 | 7 - 8 | 969 | 81 |
NM_001174117.1 | NP_001167588.1 | 7 - 8 | 969 | 81 | |
NM_015263.3 | NP_056078.2 | 7 - 8 | 969 | 81 | |
XM_005254255.1 | XP_005254312.1 | 7 - 8 | 1032 | 81 | |
XM_005254256.1 | XP_005254313.1 | 7 - 8 | 1032 | 81 | |
XM_011521402.2 | XP_011519704.1 | 7 - 8 | 1032 | 81 | |
XM_017022034.1 | XP_016877523.1 | 7 - 8 | 705 | 81 | |
GenBank mRNA | AF389880.1 | - | 7 - 8 | 951 | 81 |
BC140781.1 | - | 7 - 8 | 834 | 81 | |
BC144539.1 | - | 7 - 8 | 834 | 81 |