The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130045.1 | NP_001123517.1 | 4 - 5 | 267 | 97 |
XM_011541177.2 | XP_011539479.1 | 4 - 5 | 344 | 97 | |
XM_017000907.1 | XP_016856396.1 | 1 - 2 | 426 | 97 | |
XM_017000908.1 | XP_016856397.1 | 1 - 2 | 426 | 97 | |
XM_017000909.1 | XP_016856398.1 | 1 - 2 | 426 | 97 | |
XM_017000910.1 | XP_016856399.1 | 1 - 2 | 426 | 97 | |
XM_017000911.1 | XP_016856400.1 | 1 - 2 | 426 | 97 | |
XM_017000912.1 | XP_016856401.1 | 2 - 3 | 198 | 97 | |
GenBank mRNA | AK124125.1 | - | 4 - 5 | 267 | 97 |
BC027945.1 | - | 4 - 5 | 289 | 97 |