The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152630.4 | NP_689843.1 | 2 - 3 | 231 | 167 |
GenBank mRNA | AK313875.1 | - | 2 - 3 | 202 | 167 |
BC028710.2 | - | 2 - 3 | 200 | 167 | |
BC030784.2 | - | 2 - 3 | 212 | 167 | |
BC034979.2 | - | 2 - 3 | 197 | 167 | |
BX506299.1 | - | 2 - 3 | 233 | 167 | |
BX537938.1 | - | 2 - 3 | 233 | 167 |