The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173582.4 | NP_775853.2 | 10 - 11 | 1620 | 92 |
XM_011544953.2 | XP_011543255.1 | 11 - 12 | 1675 | 92 | |
GenBank mRNA | AB019210.1 | - | 10 - 11 | 1571 | 92 |
AK056591.1 | - | 10 - 11 | 1578 | 92 | |
BC059360.1 | - | 10 - 11 | 1584 | 92 |