Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGM2L1
Entrez Gene ID
283209
Gene Name
phosphoglucomutase 2 like 1
Gene Aliases
BM32A, PMMLP
Chromosome Location
Chr.11: 74330312 - 74398465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173582.4 NP_775853.2 11 - 12 1738 121
XM_011544953.2 XP_011543255.1 12 - 13 1793 121
GenBank mRNA AB019210.1 - 11 - 12 1689 121
AK056591.1 - 11 - 12 1696 121
BC059360.1 - 11 - 12 1702 121

View Full Product Details