The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173582.4 | NP_775853.2 | 7 - 8 | 1247 | 125 |
XM_011544953.2 | XP_011543255.1 | 8 - 9 | 1302 | 125 | |
GenBank mRNA | AB019210.1 | - | 7 - 8 | 1198 | 125 |
AK056591.1 | - | 7 - 8 | 1205 | 125 | |
BC059360.1 | - | 7 - 8 | 1211 | 125 |