The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144382.1 | NP_001137854.1 | 3 - 4 | 3102 | 112 |
NM_015184.5 | NP_055999.2 | 3 - 4 | 2896 | 112 | |
XM_006713073.3 | XP_006713136.1 | 3 - 4 | 11085 | 112 | |
XM_017006022.1 | XP_016861511.1 | 4 - 5 | 3327 | 112 | |
XM_017006023.1 | XP_016861512.1 | 4 - 5 | 3637 | 112 | |
XM_017006024.1 | XP_016861513.1 | 4 - 5 | 3256 | 112 | |
XM_017006025.1 | XP_016861514.1 | 4 - 5 | 11189 | 112 | |
GenBank mRNA | AB029015.1 | - | 3 - 4 | 3102 | 112 |
AK023546.1 | - | 2 - 3 | 1874 | 112 | |
AK291419.1 | - | 3 - 4 | 2916 | 112 | |
AL117515.1 | - | 2 - 3 | 270 | 112 | |
BC036392.2 | - | 3 - 4 | 2896 | 112 |