The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002901.2 | NP_002892.1 | 5 - 6 | 1154 | 124 |
GenBank mRNA | AK094360.1 | - | 4 - 5 | 1244 | 124 |
AK126419.1 | - | 1 - 2 | 1485 | 124 | |
AK129791.1 | - | 5 - 6 | 1181 | 124 | |
AY423728.1 | - | 3 - 4 | 390 | 124 | |
BC010120.2 | - | 5 - 6 | 1023 | 124 | |
D42073.1 | - | 5 - 6 | 940 | 124 | |
FJ224346.1 | - | 5 - 6 | 940 | 124 |