Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAS3
Entrez Gene ID
3038
Gene Name
hyaluronan synthase 3
Gene Aliases
-
Chromosome Location
Chr.16: 69105564 - 69118719 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005329.2 NP_005320.2 1 - 2 154 153
GenBank mRNA AF234839.1 - 1 - 2 154 153

View Full Product Details