The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017925.6 | NP_060395.5 | 25 - 26 | 4724 | 147 |
NR_073201.3 | 25 - 26 | 4590 | 147 | ||
XM_006716804.3 | XP_006716867.1 | 26 - 27 | 4839 | 147 | |
XM_006716805.3 | XP_006716868.1 | 26 - 27 | 4839 | 147 | |
XM_006716807.3 | XP_006716870.1 | 17 - 18 | 3364 | 147 | |
XM_017014917.1 | XP_016870406.1 | 25 - 26 | 4632 | 147 | |
XM_017014918.1 | XP_016870407.1 | 27 - 28 | 5039 | 147 | |
XM_017014919.1 | XP_016870408.1 | 27 - 28 | 5006 | 147 | |
XM_017014920.1 | XP_016870409.1 | 26 - 27 | 4863 | 147 | |
GenBank mRNA | AK000627.1 | - | 4 - 5 | 827 | 147 |
AK000693.1 | - | 7 - 8 | 927 | 147 | |
AK001046.1 | - | 2 - 3 | 305 | 147 | |
AK025705.1 | - | 5 - 6 | 1745 | 147 | |
AK125842.1 | - | 19 - 20 | 3586 | 147 | |
AK127734.1 | - | 14 - 15 | 3000 | 147 | |
AK299842.1 | - | 12 - 13 | 2645 | 147 | |
AK302901.1 | - | 7 - 8 | 2056 | 147 | |
AL834521.1 | - | 4 - 5 | 999 | 147 | |
BC144484.1 | - | 22 - 23 | 4446 | 147 | |
BC144485.1 | - | 21 - 22 | 3822 | 147 | |
CR627367.1 | - | 6 - 7 | 1930 | 147 |