The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007372.3 | NP_031398.2 | 11 - 12 | 1434 | 126 |
NM_203499.2 | NP_987095.1 | 10 - 11 | 1363 | 126 | |
XM_006721657.1 | XP_006721720.1 | 11 - 12 | 1571 | 126 | |
XM_011524259.2 | XP_011522561.1 | 5 - 6 | 1372 | 126 | |
XM_017024111.1 | XP_016879600.1 | 12 - 13 | 1670 | 126 | |
XM_017024112.1 | XP_016879601.1 | 6 - 7 | 750 | 126 | |
GenBank mRNA | AB036090.1 | - | 9 - 10 | 1159 | 126 |
AF083255.1 | - | 8 - 9 | 819 | 126 | |
AK095772.1 | - | 8 - 9 | 848 | 126 | |
AK126480.1 | - | 3 - 4 | 1461 | 126 | |
AK292908.1 | - | 8 - 9 | 946 | 126 | |
BC015505.1 | - | 9 - 10 | 1144 | 126 | |
BC078667.1 | - | 9 - 10 | 1102 | 126 | |
BC093081.1 | - | 11 - 12 | 1398 | 126 |