The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007372.3 | NP_031398.2 | 15 - 16 | 1953 | 105 |
NM_203499.2 | NP_987095.1 | 14 - 15 | 1882 | 105 | |
XM_006721657.1 | XP_006721720.1 | 15 - 16 | 2090 | 105 | |
XM_011524259.2 | XP_011522561.1 | 9 - 10 | 1891 | 105 | |
XM_017024111.1 | XP_016879600.1 | 16 - 17 | 2189 | 105 | |
XM_017024112.1 | XP_016879601.1 | 10 - 11 | 1269 | 105 | |
GenBank mRNA | AB036090.1 | - | 13 - 14 | 1678 | 105 |
AF083255.1 | - | 12 - 13 | 1338 | 105 | |
AK001964.1 | - | 1 - 2 | 163 | 105 | |
AK122737.1 | - | 1 - 2 | 932 | 105 | |
AK292908.1 | - | 12 - 13 | 1465 | 105 | |
BC015505.1 | - | 13 - 14 | 1663 | 105 | |
BC078667.1 | - | 13 - 14 | 1621 | 105 | |
BC093081.1 | - | 15 - 16 | 1917 | 105 |