The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018054.5 | NP_060524.4 | 16 - 17 | 1607 | 63 |
XM_011545878.2 | XP_011544180.1 | 17 - 18 | 1696 | 63 | |
XM_017023390.1 | XP_016878879.1 | 17 - 18 | 1696 | 63 | |
GenBank mRNA | AJ306731.1 | - | 16 - 17 | 1490 | 63 |
AK023281.1 | - | 13 - 14 | 1259 | 63 | |
AK027567.1 | - | 16 - 17 | 1560 | 63 | |
AK301500.1 | - | 14 - 15 | 1296 | 63 |