The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013321.3 | NP_037453.1 | 7 - 8 | 966 | 58 |
XM_011515329.2 | XP_011513631.1 | 7 - 8 | 1079 | 58 | |
XM_011515330.1 | XP_011513632.1 | 7 - 8 | 968 | 58 | |
XM_017012083.1 | XP_016867572.1 | 7 - 8 | 1036 | 58 | |
XM_017012084.1 | XP_016867573.1 | 7 - 8 | 1078 | 58 | |
GenBank mRNA | AF121858.1 | - | 7 - 8 | 922 | 58 |
AK074544.1 | - | 7 - 8 | 915 | 58 | |
AK315670.1 | - | 7 - 8 | 936 | 58 | |
BC007785.2 | - | 7 - 8 | 895 | 58 | |
BC021565.1 | - | 7 - 8 | 955 | 58 |