The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020717.3 | NP_065768.2 | 5 - 6 | 2980 | 111 |
NR_027121.1 | 5 - 6 | 2980 | 111 | ||
GenBank mRNA | AB033028.1 | - | 5 - 6 | 2980 | 111 |
BC151240.1 | - | 5 - 6 | 2991 | 111 |