Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 4 - 5 483 75
XM_006718397.2 XP_006718460.1 9 - 10 1328 75
XM_006718398.3 XP_006718461.1 5 - 6 1940 75
XM_011520504.1 XP_011518806.1 10 - 11 1371 75
XM_011520507.2 XP_011518809.1 9 - 10 1292 75
XM_011520508.1 XP_011518810.1 9 - 10 1328 75
XM_011520509.1 XP_011518811.1 9 - 10 1328 75
XM_011520510.1 XP_011518812.1 9 - 10 1328 75
XM_011520511.1 XP_011518813.1 9 - 10 1217 75
XM_011520512.1 XP_011518814.1 5 - 6 592 75
XM_011520513.1 XP_011518815.1 5 - 6 590 75
XM_011520514.2 XP_011518816.1 5 - 6 546 75
XM_011520515.1 XP_011518817.1 6 - 7 759 75
XM_011520516.1 XP_011518818.1 6 - 7 759 75
XM_011520517.2 XP_011518819.1 6 - 7 2081 75
XM_011520518.1 XP_011518820.1 6 - 7 729 75
XM_011520519.1 XP_011518821.1 5 - 6 617 75
XM_011520522.2 XP_011518824.1 4 - 5 1795 75
XM_011520523.2 XP_011518825.1 5 - 6 2677 75
XM_011520524.1 XP_011518826.1 9 - 10 1328 75
XM_011520525.1 XP_011518827.1 9 - 10 1328 75
XM_017018629.1 XP_016874118.1 9 - 10 1721 75
XM_017018630.1 XP_016874119.1 4 - 5 683 75
XM_017018631.1 XP_016874120.1 4 - 5 835 75
GenBank mRNA AB002296.2 - 4 - 5 483 75
AK308310.1 - 6 - 7 729 75

View Full Product Details