The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024828.3 | NP_079104.3 | 1 - 2 | 418 | 110 |
GenBank mRNA | AK023719.1 | - | 1 - 2 | 392 | 110 |
BC014658.1 | - | 1 - 2 | 394 | 110 | |
BC071953.1 | - | 1 - 2 | 317 | 110 | |
BP228742.1 | - | 1 - 2 | 418 | 110 |