The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289396.1 | NP_001276325.1 | 16 - 17 | 2625 | 88 |
NM_001289397.1 | NP_001276326.1 | 16 - 17 | 2638 | 88 | |
NM_003070.4 | NP_003061.3 | 16 - 17 | 2638 | 88 | |
NM_139045.3 | NP_620614.2 | 16 - 17 | 2638 | 88 | |
GenBank mRNA | AK296373.1 | - | 16 - 17 | 2026 | 88 |
AK299683.1 | - | 7 - 8 | 831 | 88 | |
X72889.1 | - | 16 - 17 | 2626 | 88 |