The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005622.3 | NP_005613.2 | 9 - 10 | 1414 | 108 |
XM_011545911.1 | XP_011544213.1 | 10 - 11 | 1519 | 108 | |
XM_017023523.1 | XP_016879012.1 | 9 - 10 | 1408 | 108 | |
XM_017023524.1 | XP_016879013.1 | 8 - 9 | 1278 | 108 | |
GenBank mRNA | AK290322.1 | - | 9 - 10 | 1383 | 108 |
AK296122.1 | - | 9 - 10 | 1213 | 108 | |
BI826935.1 | - | 3 - 4 | 289 | 108 | |
BU170035.1 | - | 3 - 4 | 266 | 108 | |
D16350.1 | - | 8 - 9 | 1254 | 108 |