Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
-
Chromosome Location
Chr.10: 101845599 - 102056237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024541.2 NP_078817.2 11 - 12 936 84
XM_006717969.1 XP_006718032.1 11 - 12 934 84
XM_011540151.2 XP_011538453.1 11 - 12 981 84
XM_011540152.2 XP_011538454.1 11 - 12 934 84
XM_011540153.2 XP_011538455.1 11 - 12 934 84
XM_011540154.2 XP_011538456.1 11 - 12 934 84
XM_011540156.2 XP_011538458.1 4 - 5 900 84
XM_011540157.2 XP_011538459.1 10 - 11 885 84
XM_017016635.1 XP_016872124.1 11 - 12 934 84
XM_017016636.1 XP_016872125.1 11 - 12 934 84
GenBank mRNA DT932690.1 - 3 - 4 84

View Full Product Details