The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286778.1 | NP_001273707.1 | 4 - 5 | 624 | 72 |
NM_031461.5 | NP_113649.1 | 5 - 6 | 1136 | 72 | |
GenBank mRNA | AF142573.1 | - | 5 - 6 | 732 | 72 |
AF329197.1 | - | 4 - 5 | 711 | 72 | |
AK304034.1 | - | 4 - 5 | 612 | 72 | |
AK314048.1 | - | 5 - 6 | 892 | 72 | |
AL834301.1 | - | 5 - 6 | 868 | 72 | |
AY358564.1 | - | 5 - 6 | 1094 | 72 | |
BC020514.1 | - | 5 - 6 | 1001 | 72 |