The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105518.1 | NP_001098988.1 | 9 - 10 | 1543 | 100 |
NM_017673.6 | NP_060143.4 | 9 - 10 | 1594 | 100 | |
XM_005245271.1 | XP_005245328.1 | 9 - 10 | 1601 | 100 | |
XM_005245273.1 | XP_005245330.1 | 9 - 10 | 1502 | 100 | |
XM_011509657.1 | XP_011507959.1 | 9 - 10 | 1581 | 100 | |
XM_017001527.1 | XP_016857016.1 | 9 - 10 | 1481 | 100 | |
XM_017001528.1 | XP_016857017.1 | 9 - 10 | 1601 | 100 | |
XM_017001529.1 | XP_016857018.1 | 10 - 11 | 1806 | 100 | |
XM_017001530.1 | XP_016857019.1 | 9 - 10 | 1601 | 100 | |
GenBank mRNA | AF288392.1 | - | 8 - 9 | 1440 | 100 |
AK093263.1 | - | 3 - 4 | 306 | 100 | |
AK127980.1 | - | 9 - 10 | 1481 | 100 | |
AK291443.1 | - | 9 - 10 | 1460 | 100 | |
AK315802.1 | - | 9 - 10 | 1561 | 100 | |
BC030781.2 | - | 9 - 10 | 1596 | 100 |