The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105518.1 | NP_001098988.1 | 16 - 17 | 2553 | 149 |
NM_017673.6 | NP_060143.4 | 16 - 17 | 2604 | 149 | |
XM_005245271.1 | XP_005245328.1 | 16 - 17 | 2611 | 149 | |
XM_005245273.1 | XP_005245330.1 | 16 - 17 | 2512 | 149 | |
XM_011509657.1 | XP_011507959.1 | 16 - 17 | 2591 | 149 | |
XM_017001527.1 | XP_016857016.1 | 16 - 17 | 2491 | 149 | |
XM_017001529.1 | XP_016857018.1 | 17 - 18 | 2816 | 149 | |
GenBank mRNA | AF288392.1 | - | 15 - 16 | 2450 | 149 |
AK000128.1 | - | 4 - 5 | 603 | 149 | |
AK093263.1 | - | 10 - 11 | 1316 | 149 | |
AK127980.1 | - | 16 - 17 | 2491 | 149 | |
AK225238.1 | - | 4 - 5 | 603 | 149 | |
AK291443.1 | - | 16 - 17 | 2470 | 149 | |
AK315802.1 | - | 16 - 17 | 2571 | 149 | |
BC030781.2 | - | 16 - 17 | 2606 | 149 |