The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105518.1 | NP_001098988.1 | 17 - 18 | 2618 | 146 |
NM_017673.6 | NP_060143.4 | 17 - 18 | 2669 | 146 | |
XM_005245271.1 | XP_005245328.1 | 17 - 18 | 2676 | 146 | |
XM_005245273.1 | XP_005245330.1 | 17 - 18 | 2577 | 146 | |
XM_011509657.1 | XP_011507959.1 | 17 - 18 | 2656 | 146 | |
XM_017001527.1 | XP_016857016.1 | 17 - 18 | 2556 | 146 | |
XM_017001529.1 | XP_016857018.1 | 18 - 19 | 2881 | 146 | |
GenBank mRNA | AF288392.1 | - | 16 - 17 | 2515 | 146 |
AK000128.1 | - | 5 - 6 | 668 | 146 | |
AK093263.1 | - | 11 - 12 | 1381 | 146 | |
AK127980.1 | - | 17 - 18 | 2556 | 146 | |
AK225238.1 | - | 5 - 6 | 668 | 146 | |
AK291443.1 | - | 17 - 18 | 2535 | 146 | |
AK315802.1 | - | 17 - 18 | 2636 | 146 | |
BC030781.2 | - | 17 - 18 | 2671 | 146 |