Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RENBP
Entrez Gene ID
5973
Gene Name
renin binding protein
Gene Aliases
RBP, RNBP
Chromosome Location
Chr.X: 153935263 - 153944780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002910.5 NP_002901.2 3 - 4 381 71
XM_017029698.1 XP_016885187.1 3 - 4 276 71
GenBank mRNA AK298125.1 - 3 - 4 244 71
BC015558.2 - 3 - 4 224 71
D10232.1 - 3 - 4 211 71

View Full Product Details