The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002910.5 | NP_002901.2 | 4 - 5 | 457 | 99 |
XM_017029698.1 | XP_016885187.1 | 4 - 5 | 352 | 99 | |
GenBank mRNA | AK298125.1 | - | 4 - 5 | 320 | 99 |
BC015558.2 | - | 4 - 5 | 300 | 99 | |
D10232.1 | - | 4 - 5 | 287 | 99 |