The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130958.1 | NP_001124430.1 | 1 - 2 | 86 | 74 |
GenBank mRNA | DQ132786.1 | - | 1 - 2 | 87 | 74 |