Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 17 - 18 2897 89
XM_005266299.2 XP_005266356.1 17 - 18 2921 89
XM_006719779.2 XP_006719842.1 17 - 18 2921 89
XM_006719780.2 XP_006719843.1 17 - 18 2921 89
XM_006719781.2 XP_006719844.1 17 - 18 2921 89
XM_006719782.2 XP_006719845.1 17 - 18 2921 89
XM_006719783.2 XP_006719846.1 17 - 18 2921 89
XM_006719785.2 XP_006719848.1 17 - 18 2921 89
XM_006719786.2 XP_006719849.1 17 - 18 2921 89
XM_006719787.2 XP_006719850.1 17 - 18 2921 89
XM_006719788.2 XP_006719851.1 17 - 18 2921 89
XM_006719789.2 XP_006719852.1 17 - 18 2921 89
XM_011535004.2 XP_011533306.1 16 - 17 2460 89
XM_017020454.1 XP_016875943.1 17 - 18 2921 89
XM_017020455.1 XP_016875944.1 17 - 18 2921 89
XM_017020456.1 XP_016875945.1 17 - 18 2921 89
XM_017020457.1 XP_016875946.1 17 - 18 2921 89
XM_017020460.1 XP_016875949.1 17 - 18 2921 89
XM_017020461.1 XP_016875950.1 17 - 18 2921 89
XM_017020462.1 XP_016875951.1 17 - 18 2921 89
XM_017020464.1 XP_016875953.1 17 - 18 2921 89
XM_017020466.1 XP_016875955.1 17 - 18 2921 89
XM_017020467.1 XP_016875956.1 17 - 18 2921 89
XM_017020468.1 XP_016875957.1 16 - 17 2408 89
GenBank mRNA AF075587.1 - 18 - 19 2662 89

View Full Product Details