Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 21 - 22 3325 79
XM_005266299.2 XP_005266356.1 21 - 22 3349 79
XM_006719779.2 XP_006719842.1 21 - 22 3349 79
XM_006719780.2 XP_006719843.1 21 - 22 3349 79
XM_006719781.2 XP_006719844.1 21 - 22 3349 79
XM_006719782.2 XP_006719845.1 21 - 22 3349 79
XM_006719783.2 XP_006719846.1 21 - 22 3349 79
XM_006719784.2 XP_006719847.1 20 - 21 3241 79
XM_006719785.2 XP_006719848.1 21 - 22 3349 79
XM_006719786.2 XP_006719849.1 21 - 22 3349 79
XM_006719787.2 XP_006719850.1 21 - 22 3349 79
XM_006719788.2 XP_006719851.1 21 - 22 3349 79
XM_006719789.2 XP_006719852.1 21 - 22 3349 79
XM_011535004.2 XP_011533306.1 20 - 21 2888 79
XM_011535005.2 XP_011533307.1 4 - 5 429 79
XM_017020454.1 XP_016875943.1 21 - 22 3349 79
XM_017020455.1 XP_016875944.1 21 - 22 3349 79
XM_017020456.1 XP_016875945.1 21 - 22 3349 79
XM_017020457.1 XP_016875946.1 21 - 22 3349 79
XM_017020458.1 XP_016875947.1 20 - 21 3241 79
XM_017020459.1 XP_016875948.1 20 - 21 3241 79
XM_017020460.1 XP_016875949.1 21 - 22 3349 79
XM_017020461.1 XP_016875950.1 21 - 22 3349 79
XM_017020462.1 XP_016875951.1 21 - 22 3349 79
XM_017020463.1 XP_016875952.1 20 - 21 3241 79
XM_017020464.1 XP_016875953.1 21 - 22 3349 79
XM_017020465.1 XP_016875954.1 20 - 21 3241 79
XM_017020466.1 XP_016875955.1 21 - 22 3349 79
XM_017020467.1 XP_016875956.1 21 - 22 3349 79
XM_017020468.1 XP_016875957.1 20 - 21 2836 79
GenBank mRNA AF075587.1 - 22 - 23 3090 79
BX473679.1 - 4 - 5 391 79

View Full Product Details